siRNA Details
| Gene Symbol: CDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139286.3 | NP_644815.1 | 4 | 549 | ||
| XM_017014573.1 | XP_016870062.1 | 4 | 516 | |||
| XM_017014574.1 | XP_016870063.1 | 4 | 981 | |||
| XM_017014575.1 | XP_016870064.1 | 4 | 511 | |||
| GenBank mRNA | AF503918.1 | - | 1 | 190 | ||
| BC042534.1 | - | 4 | 504 | 4751173 | ||
| BC066300.1 | - | 4 | 534 | 5258904 | ||
| BF244254.1 | - | 4 | 547 | |||

CDC26
Genomic Map