siRNA Details
| Gene Symbol: NOL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006722563.2 | XP_006722626.1 | 1 | 735 | ||
| XM_011526237.1 | XP_011524539.1 | 1 | 735 | |||
| XM_011526238.1 | XP_011524540.1 | 1 | 735 | |||
| XM_017026055.1 | XP_016881544.1 | 1 | 735 | |||
| XM_017026059.1 | XP_016881548.1 | 1 | 734 | |||

NOL4
Genomic Map