siRNA Details
| Gene Symbol: UBE2Q2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243531.1 | NP_001230460.1 | 3 | 335 | ||
| XM_017021854.1 | XP_016877343.1 | 3 | 895 | |||
| XM_017021855.1 | XP_016877344.1 | 3 | 892 | |||
| GenBank mRNA | CB050071.1 | - | 3 | 341 | ||

UBE2Q2L
Genomic Map