siRNA Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127181.2 | NP_001120653.1 | 1 | 251 | ||
| NM_033319.3 | NP_201576.1 | 1 | 251 | |||
| GenBank mRNA | AK056348.1 | - | 1 | 437 | ||
| DA472686.1 | - | 1 | 492 | |||
| DB244459.1 | - | 1 | 500 | |||
| DN995099.1 | - | 1 | 259 | |||

CENPL
Genomic Map