siRNA Details
| Gene Symbol: LOC101930010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125866.1 | 1 | 24 | |||
| GenBank mRNA | CK819990.1 | - | 1 | 24 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101930010| Gene Symbol: LOC101930010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125866.1 | 1 | 24 | |||
| GenBank mRNA | CK819990.1 | - | 1 | 24 | ||