siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011545916.2 | XP_011544218.1 | 1 | 1252 | ||
| XM_011545917.2 | XP_011544219.1 | 1 | 1252 | |||
| XM_011545919.2 | XP_011544221.1 | 1 | 1251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBRS| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011545916.2 | XP_011544218.1 | 1 | 1252 | ||
| XM_011545917.2 | XP_011544219.1 | 1 | 1252 | |||
| XM_011545919.2 | XP_011544221.1 | 1 | 1251 | |||