siRNA Details
| Gene Symbol: MEIS1-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046438.2 | 3, 4 | 277 | |||
| GenBank mRNA | DA226783.1 | - | 3, 4 | 277 | ||
| DA702499.1 | - | 2, 3 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIS1-AS3| Gene Symbol: MEIS1-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046438.2 | 3, 4 | 277 | |||
| GenBank mRNA | DA226783.1 | - | 3, 4 | 277 | ||
| DA702499.1 | - | 2, 3 | 75 | |||