siRNA Details
| Gene Symbol: LOC105371303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003020.1 | XP_016858509.1 | 4 | 759 | ||
| Gene Symbol: NBPF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK308678.1 | - | 2 | 514 | ||

LOC105371303
Genomic Map