siRNA Details
| Gene Symbol: SSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093431.1 | - | 2 | 237 | ||
| AK123170.1 | - | 7 | 969 | |||
| AK124542.1 | - | 1 | 838 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSPO| Gene Symbol: SSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093431.1 | - | 2 | 237 | ||
| AK123170.1 | - | 7 | 969 | |||
| AK124542.1 | - | 1 | 838 | |||