siRNA Details
| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284261.1 | NP_001271190.1 | 1 | 15 | ||
| NM_001284262.1 | NP_001271191.1 | 1 | 15 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR86| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284261.1 | NP_001271190.1 | 1 | 15 | ||
| NM_001284262.1 | NP_001271191.1 | 1 | 15 | |||