siRNA Details
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 1 | 22 | ||
NM_033285.3 | NP_150601.1 | 1 | 22 | |||
GenBank mRNA | DB086880.1 | - | 1 | 22 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 1 | 22 | ||
NM_033285.3 | NP_150601.1 | 1 | 22 | |||
GenBank mRNA | DB086880.1 | - | 1 | 22 |