siRNA Details
| Gene Symbol: UBE2S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014501.2 | NP_055316.2 | 4 | 1005 | ||
| GenBank mRNA | AB062397.1 | - | 1 | 815 | ||
| BC004236.2 | - | 1 | 815 | 2988449 | ||
| BC007554.1 | - | 1 | 815 | 2967547 | ||
| BC065364.1 | - | 1 | 825 | 5434942 | ||
| BC066948.1 | - | 1 | 661 | 4836851 | ||
| M91670.1 | - | 4 | 696 | |||

UBE2S
Genomic Map