siRNA Details
| Gene Symbol: RABEPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174152.1 | NP_001167623.1 | 9 | 1124 | ||
| NM_001174153.1 | NP_001167624.1 | 8 | 971 | |||
| NM_005833.3 | NP_005824.2 | 8 | 1285 | |||
| XM_005251640.4 | XP_005251697.1 | 8 | 1197 | |||
| XM_005251641.4 | XP_005251698.1 | 7 | 1052 | |||
| XM_005251642.4 | XP_005251699.1 | 7 | 1043 | |||
| XM_005251644.4 | XP_005251701.1 | 6 | 893 | |||
| XM_011518120.2 | XP_011516422.1 | 8 | 1359 | |||
| XM_017014177.1 | XP_016869666.1 | 9 | 1109 | |||
| XM_017014178.1 | XP_016869667.1 | 8 | 1011 | |||
| XM_017014179.1 | XP_016869668.1 | 7 | 1084 | |||
| XM_017014180.1 | XP_016869669.1 | 7 | 858 | |||
| XM_017014181.1 | XP_016869670.1 | 6 | 969 | |||
| XM_017014182.1 | XP_016869671.1 | 7 | 794 | |||
| XM_017014183.1 | XP_016869672.1 | 7 | 1290 | |||
| GenBank mRNA | AK290768.1 | - | 8 | 1129 | ||
| BC000503.2 | - | 8 | 1262 | 2821471 | ||
| BC047023.1 | - | 8 | 951 | 5396980 | ||
| BC053541.1 | - | 8 | 1094 | 5724458 | ||
| BC065725.1 | - | 9 | 1127 | 4102355 | ||
| CR456837.1 | - | 7 | 955 | |||
| CR541851.1 | - | 7 | 955 | |||
| KU178507.1 | - | 7 | 955 | |||
| KU178508.1 | - | 7 | 868 | |||
| KU178509.1 | - | 6 | 806 | |||
| KU178510.1 | - | 6 | 802 | |||
| KU178511.1 | - | 5 | 640 | |||
| Z97074.1 | - | 9 | 1104 | |||

RABEPK
Genomic Map