siRNA Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167947.2 | NP_001161419.1 | 7 | 717 | ||
| NM_005834.4 | NP_005825.1 | 6 | 567 | |||
| GenBank mRNA | AF034790.1 | - | 6 | 567 | ||
| AF077039.1 | - | 5 | 432 | |||
| AJ005895.1 | - | 5 | 420 | |||
| AK290207.1 | - | 5 | 442 | |||
| BC010142.2 | - | 5 | 421 | 4300787 | ||
| CB993524.1 | - | 6 | 608 | |||

TIMM17B
Genomic Map