siRNA Details
| Gene Symbol: PLEKHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005259256.3 | XP_005259313.1 | 1 | 361 | ||
| GenBank mRNA | AK225814.1 | - | 1 | 305 | ||
| AK301616.1 | - | 1 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHF1| Gene Symbol: PLEKHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005259256.3 | XP_005259313.1 | 1 | 361 | ||
| GenBank mRNA | AK225814.1 | - | 1 | 305 | ||
| AK301616.1 | - | 1 | 325 | |||