siRNA Details
| Gene Symbol: ZDHHC11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010116.1 | XP_016865605.1 | 14 | 4484 | ||
| XM_017010120.1 | XP_016865609.1 | 15 | 5432 | |||
| XM_017010122.1 | XP_016865611.1 | 15 | 5429 | |||
| Gene Symbol: ZDHHC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096220.1 | - | 2204 | |||

ZDHHC11B
Genomic Map