siRNA Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030930.3 | NP_112192.2 | 11 | 1924 | ||
| XM_011545290.1 | XP_011543592.1 | 9 | 1534 | |||
| XM_011545291.2 | XP_011543593.1 | 10 | 1816 | |||
| GenBank mRNA | AJ271326.1 | - | 11 | 1883 | ||
| AY007125.1 | - | 10 | 1711 | |||
| BC092472.1 | - | 11 | 1892 | 30348121 | ||

UNC93B1
Genomic Map