siRNA Details
| Gene Symbol: ZNF282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303481.1 | NP_001290410.1 | 9 | 3420 | ||
| NM_003575.3 | NP_003566.1 | 8 | 3677 | |||
| XM_006716151.3 | XP_006716214.1 | 8 | 3674 | |||
| GenBank mRNA | BC009542.1 | - | 1 | 520 | ||
| BC041704.1 | - | 8 | 3404 | |||
| D30612.1 | - | 8 | 3681 | |||

ZNF282
Genomic Map