siRNA Details
| Gene Symbol: LOC732272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092688.1 | - | 5, 6 | 1134 | ||
| Gene Symbol: ZNF429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057495.1 | - | 6, 7 | 1114 | ||

LOC732272
Genomic Map