siRNA Details
| Gene Symbol: HCG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028032.1 | 1 | 352 | |||
| GenBank mRNA | AK289926.1 | - | 1 | 375 | ||
| DA293452.1 | - | 1 | 375 | |||
| X95289.1 | - | 1 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG9| Gene Symbol: HCG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028032.1 | 1 | 352 | |||
| GenBank mRNA | AK289926.1 | - | 1 | 375 | ||
| DA293452.1 | - | 1 | 375 | |||
| X95289.1 | - | 1 | 352 | |||