siRNA Details
| Gene Symbol: UQCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085372.2 | NP_001078841.1 | 1, 2 | 204 | ||
| GenBank mRNA | AY358935.1 | - | 1, 2 | 204 | ||
| BC090057.1 | - | 1, 2 | 172 | 30347387 | ||
| BG385605.1 | - | 1, 2 | 176 | |||
| CF131173.1 | - | 1, 2 | 476 | |||

UQCC3
Genomic Map