siRNA Details
| Gene Symbol: SCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007098.2 | NP_001007099.1 | 5, 6 | 568 | ||
| NM_001193599.1 | NP_001180528.1 | 5, 6 | 628 | |||
| NM_001193600.1 | NP_001180529.1 | 5, 6 | 568 | |||
| NM_001193617.1 | NP_001180546.1 | 5, 6 | 642 | |||
| NM_002979.4 | NP_002970.2 | 6, 7 | 700 | |||
| XM_005271103.4 | XP_005271160.1 | 6, 7 | 643 | |||
| XM_011541935.2 | XP_011540237.1 | 6, 7 | 643 | |||
| XM_017002045.1 | XP_016857534.1 | 6, 7 | 643 | |||
| GenBank mRNA | AB208789.1 | - | 6, 7 | 652 | ||
| AK294631.1 | - | 5, 6 | 571 | |||
| AK295214.1 | - | 5, 6 | 585 | |||
| AK308105.1 | - | 5, 6 | 511 | |||
| AK312856.1 | - | 6, 7 | 651 | |||
| BC067108.1 | - | 5, 6 | 479 | 5176900 | ||
| M75883.1 | - | 6, 7 | 553 | |||

SCP2
Genomic Map