siRNA Details
| Gene Symbol: GPR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195381.1 | NP_001182310.1 | 3 | 493 | ||
| NM_001195382.1 | NP_001182311.1 | 3 | 493 | |||
| GenBank mRNA | AK172786.1 | - | 3 | 470 | ||
| AK290256.1 | - | 3 | 456 | |||
| DB011311.1 | - | 3 | 493 | |||

GPR35
Genomic Map