siRNA Details
| Gene Symbol: CRLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012288.2 | NP_001012288.2 | 4 | 525 | ||
| NM_022148.3 | NP_071431.2 | 5 | 628 | |||
| XM_011546181.2 | XP_011544483.1 | 5 | 663 | |||
| GenBank mRNA | AB052639.1 | - | 5 | 539 | ||
| AF142570.1 | - | 5 | 555 | |||
| AF338733.1 | - | 5 | 539 | |||
| AK223303.1 | - | 5 | 655 | |||
| AY775789.1 | - | 4 | 436 | |||

CRLF2
Genomic Map