siRNA Details
| Gene Symbol: HIST2H3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005464.2 | NP_001005464.1 | 1 | 397 | ||
| Gene Symbol: HIST2H3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021059.2 | NP_066403.2 | 1 | 397 | ||
| GenBank mRNA | BC015544.1 | - | 1 | 397 | ||
| BC074969.2 | - | 1 | 389 | 30915509 | ||
| BC130635.1 | - | 1 | 391 | 40146666 | ||
| BC130637.1 | - | 1 | 391 | 40146668 | ||

HIST2H3A
Genomic Map