siRNA Details
| Gene Symbol: CTAGE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178561.4 | NP_848656.2 | 1 | 1288 | ||
| GenBank mRNA | BC101323.1 | - | 1 | 546 | ||
| BC101324.2 | - | 1 | 546 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 CTAGE6
CTAGE6| Gene Symbol: CTAGE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178561.4 | NP_848656.2 | 1 | 1288 | ||
| GenBank mRNA | BC101323.1 | - | 1 | 546 | ||
| BC101324.2 | - | 1 | 546 | |||
 Back To Top
 Back To Top