siRNA Details
| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006382.3 | NP_006373.2 | 12 | 2242 | ||
| GenBank mRNA | BC143291.1 | - | 5 | 518 | ||
| BC171723.1 | - | 3 | 280 | |||
| U43383.1 | - | 5 | 536 | |||
| U65652.1 | - | 5 | 771 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDRT1| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006382.3 | NP_006373.2 | 12 | 2242 | ||
| GenBank mRNA | BC143291.1 | - | 5 | 518 | ||
| BC171723.1 | - | 3 | 280 | |||
| U43383.1 | - | 5 | 536 | |||
| U65652.1 | - | 5 | 771 | |||