siRNA Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005855.3 | NP_005846.1 | 1, 2 | 192 | ||
| XM_017003155.1 | XP_016858644.1 | 1, 2 | 120 | |||
| GenBank mRNA | AI017108.1 | - | 1, 2 | 192 | ||
| AJ001014.1 | - | 1, 2 | 92 | |||
| AY265457.1 | - | 1, 2 | 60 | |||
| BC000548.2 | - | 1, 2 | 84 | 3163522 | ||
| CR542032.1 | - | 1, 2 | 60 | |||
| CR542044.1 | - | 1, 2 | 60 | |||

RAMP1
Genomic Map