siRNA Details
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 10 | 1858 | |||
GenBank mRNA | BC018097.1 | - | 8 | 1047 | ||
BC044610.1 | - | 9 | 1301 | |||
BC048987.1 | - | 10 | 1918 | |||
DQ309039.1 | - | 8 | 1037 | |||
DQ309040.1 | - | 10 | 1858 |
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026589.1 | 9 | 1747 | |||
GenBank mRNA | AK091254.1 | - | 4 | 434 | ||
BC066982.1 | - | 10 | 1598 |
