siRNA Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005858.3 | NP_005849.1 | 6 | 991 | ||
| XM_011527624.2 | XP_011525926.1 | 6 | 1084 | |||
| XM_017026141.1 | XP_016881630.1 | 6 | 1136 | |||
| GenBank mRNA | AK303640.1 | - | 3 | 618 | ||
| AK308251.1 | - | 4 | 727 | |||
| AK309529.1 | - | 2 | 361 | |||
| BC037270.2 | - | 6 | 981 | 4837309 | ||
| Y11997.1 | - | 6 | 982 | |||

AKAP8
Genomic Map