siRNA Details
| Gene Symbol: Snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133924.3 | NP_598685.2 | 3 | 1431 | ||
| GenBank mRNA | AK141663.1 | - | 3 | 1432 | ||
| AK161207.1 | - | 1 | 1030 | |||
| BC017623.1 | - | 1 | 478 | |||
| BC037155.1 | - | 1 | 367 | |||
| BC094371.1 | - | 2 | 2127 | |||

Snx21
Genomic Map