siRNA Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198102.2 | NP_932770.2 | 4 | 580 | ||
| GenBank mRNA | AK089980.1 | - | 4 | 559 | ||
| AK171708.1 | - | 4 | 562 | |||
| BC057448.1 | - | 4 | 538 | 4016620 | ||
| BC058764.1 | - | 5 | 845 | |||

Tra2a
Genomic Map