siRNA Details
| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181854.2 | NP_862902.1 | 2 | 2473 | ||
| GenBank mRNA | AK129444.1 | - | 2 | 2373 | ||
| AK132164.1 | - | 2 | 2436 | |||
| AK133990.1 | - | 2 | 2399 | |||
| AK170835.1 | - | 2 | 2416 | |||
| BC028991.1 | - | 2 | 2385 | 5355869 | ||

Champ1
Genomic Map