siRNA Details
| Gene Symbol: Enkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198299.1 | NP_938041.1 | 6 | 902 | ||
| GenBank mRNA | AK087475.1 | - | 5 | 1065 | ||
| BC018168.1 | - | 5 | 1188 | |||
| BC023921.1 | - | 5 | 1291 | |||
| BC026993.1 | - | 3 | 2969 | |||
| BC052498.1 | - | 6 | 902 | 1245862 | ||

Enkd1
Genomic Map