siRNA Details
| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042591.1 | NP_001036056.1 | 1 | 409 | ||
| GenBank mRNA | AK084951.1 | - | 1 | 420 | ||
| AK088288.1 | - | 1 | 424 | |||
| AK122499.1 | - | 1 | 396 | |||
| BC044762.1 | - | 1 | 407 | |||
| BC054844.1 | - | 1 | 409 | 6493441 | ||
| BC098474.1 | - | 1 | 403 | 30094818 | ||
| DQ861994.1 | - | 1 | 154 | |||

Arrdc3
Genomic Map