siRNA Details
| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042591.1 | NP_001036056.1 | 1, 2 | 509 | ||
| GenBank mRNA | AK084951.1 | - | 1, 2 | 520 | ||
| AK088288.1 | - | 1, 2 | 524 | |||
| AK122499.1 | - | 1, 2 | 496 | |||
| BC044762.1 | - | 1, 2 | 507 | |||
| BC054844.1 | - | 1, 2 | 509 | 6493441 | ||
| BC098474.1 | - | 1, 2 | 503 | 30094818 | ||
| DQ861994.1 | - | 1, 2 | 254 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001007797, AY383703.

Arrdc3
Genomic Map