siRNA Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285788.1 | NP_001272717.1 | 3 | 454 | ||
| NM_001285791.1 | NP_001272720.1 | 3 | 454 | |||
| NM_134109.2 | NP_598870.1 | 3 | 454 | |||
| GenBank mRNA | AK035504.1 | - | 3 | 383 | ||
| AK036844.1 | - | 3 | 380 | |||
| AK050806.1 | - | 3 | 400 | |||
| AK136284.1 | - | 3 | 454 | |||
| BC010485.1 | - | 3 | 394 | 3483341 | ||
| BC057644.1 | - | 3 | 451 | 4912789 | ||

Ildr1
Genomic Map