siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 16 | 2773 | ||
GenBank mRNA | AK044527.1 | - | 6 | 1009 | ||
AK078556.1 | - | 1 | 212 | |||
AY237726.1 | - | 16 | 2802 | |||
AY237727.2 | - | 1 | 326 | |||
BC030478.1 | - | 6 | 1005 | |||
BC045197.1 | - | 1 | 256 | |||
BC058610.1 | - | 16 | 2785 | 30056142 | ||
BC145279.1 | - | 16 | 2760 | 9053833 | ||
BC150749.1 | - | 16 | 2760 | 9087660 | ||
BC171954.1 | - | 16 | 2761 |
