siRNA Details
| Gene Symbol: Mylk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139300.3 | NP_647461.3 | 23, 24 | 4469 | ||
| GenBank mRNA | AK162008.1 | - | 5, 6 | 832 | ||
| AY237726.1 | - | 23, 24 | 4498 | |||
| AY237727.2 | - | 8, 9 | 2022 | |||
| BC030478.1 | - | 13, 14 | 2701 | |||
| BC037721.1 | - | 8, 9 | 1032 | |||
| BC045197.1 | - | 8, 9 | 1952 | |||
| BC058610.1 | - | 23, 24 | 4481 | 30056142 | ||
| BC145279.1 | - | 23, 24 | 4456 | 9053833 | ||
| BC150749.1 | - | 23, 24 | 4456 | 9087660 | ||
| BC171954.1 | - | 23, 24 | 4457 | |||

Mylk
Genomic Map