siRNA Details
| Gene Symbol: Mthfs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026829.2 | NP_081105.1 | 3 | 420 | ||
| GenBank mRNA | AK004092.1 | - | 3 | 421 | ||
| AK154942.1 | - | 4 | 1093 | |||
| AK158264.1 | - | 429 | ||||
| Gene Symbol: Mthfsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128601.1 | NP_001122073.1 | 3 | 438 | ||
| GenBank mRNA | AK009422.1 | - | 2 | 130 | ||
| AK081946.1 | - | 114 | ||||
| BC024567.1 | - | 3 | 330 | 5026828 | ||

Mthfs
Genomic Map