siRNA Details
| Gene Symbol: C330006A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 1635 | ||
| NM_001256522.1 | NP_001243451.1 | 2 | 1475 | |||
| GenBank mRNA | AK077073.1 | - | 1 | 654 | ||
| AK082757.1 | - | 1 | 657 | |||
| BC020361.1 | - | 1 | 595 | |||

C330006A16Rik
Genomic Map