siRNA Details
| Gene Symbol: Upk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026815.2 | NP_081091.1 | 8 | 1063 | ||
| GenBank mRNA | AK004014.1 | - | 8 | 1064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upk1a| Gene Symbol: Upk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026815.2 | NP_081091.1 | 8 | 1063 | ||
| GenBank mRNA | AK004014.1 | - | 8 | 1064 | ||