siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253712.1 | NP_001240641.1 | 6 | 823 | ||
| NM_001253714.1 | NP_001240643.1 | 5 | 740 | |||
| NM_001253715.1 | NP_001240644.1 | 5 | 689 | |||
| NM_001253716.1 | NP_001240645.1 | 6 | 551 | |||
| NM_026322.4 | NP_080598.2 | 11 | 1222 | |||
| GenBank mRNA | AK002356.1 | - | 6 | 698 | ||
| AK009822.1 | - | 6 | 700 | |||
| AK018338.1 | - | 6 | 545 | |||
| AK049714.1 | - | 6 | 824 | |||
| BC014738.1 | - | 5 | 648 | 4239349 | ||
| BC089311.1 | - | 11 | 1222 | 6412625 | ||
| CB953198.1 | - | 5 | 611 | |||

Msra
Genomic Map