siRNA Details
| Gene Symbol: Scgb1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009596.1 | NP_033726.1 | 2 | 185 | ||
| GenBank mRNA | BC016132.1 | - | 2 | 185 | 4922791 | |
| BC132436.1 | - | 2 | 269 | 40130713 | ||
| GU269231.1 | - | 2 | 184 | |||
| Gene Symbol: Scgb1b30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099330.2 | NP_001092800.1 | 2 | 267 | ||

Scgb1b27
Genomic Map