siRNA Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013456.2 | NP_038484.1 | 3, 4 | 440 | ||
| GenBank mRNA | AF093775.1 | - | 3, 4 | 415 | ||
| BC111890.1 | - | 3, 4 | 406 | 40060598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actn3| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013456.2 | NP_038484.1 | 3, 4 | 440 | ||
| GenBank mRNA | AF093775.1 | - | 3, 4 | 415 | ||
| BC111890.1 | - | 3, 4 | 406 | 40060598 | ||