siRNA Details
| Gene Symbol: Pop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026398.4 | NP_080674.1 | 6 | 838 | ||
| GenBank mRNA | AK005290.1 | - | 5 | 594 | ||
| AK012523.1 | - | 596 | ||||
| AK013257.1 | - | 4 | 575 | |||
| BC022670.1 | - | 6 | 862 | 4224973 | ||

Pop5
Genomic Map