siRNA Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054097.3 | NP_473438.1 | 4, 5 | 620 | ||
| GenBank mRNA | AB054591.1 | - | 4, 5 | 563 | ||
| AK148299.1 | - | 4, 5 | 580 | |||
| AK181991.1 | - | 1, 2 | 31 | |||
| AK207886.1 | - | 2, 3 | 228 | |||
| AK208052.1 | - | 1, 2 | 33 | |||
| BC021383.1 | - | 4, 5 | 553 | 5042933 | ||

Pip4k2c
Genomic Map