siRNA Details
| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080443.2 | NP_536691.2 | 4 | 914 | ||
| GenBank mRNA | AF398968.1 | - | 1 | 199 | ||
| AK051017.1 | - | 5 | 902 | |||
| BC062948.1 | - | 1 | 143 | 30532932 | ||
| BC066824.1 | - | 3 | 416 | 30087227 | ||
| BC085155.1 | - | 4 | 894 | |||

Asb7
Genomic Map