siRNA Details
| Gene Symbol: Apoe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019319.1 | - | 1 | 666 | ||
| AK131624.1 | - | 4 | 931 | |||
| AK134921.1 | - | 4 | 931 | |||
| AK149568.1 | - | 4 | 913 | |||
| AK150834.1 | - | 4 | 933 | |||
| AK159105.1 | - | 4 | 931 | |||
| AK159424.1 | - | 4 | 933 | |||
| AK159517.1 | - | 4 | 931 | |||
| AK197774.1 | - | 1 | 58 | |||
| BC028816.1 | - | 4 | 910 | 5136415 | ||
| BC044785.1 | - | 3 | 902 | |||
| BC083351.1 | - | 3 | 900 | 6818357 | ||
| CT010212.1 | - | 3 | 865 | |||
| CT010356.1 | - | 3 | 865 | |||
| M12414.1 | - | 3 | 865 | |||
| M73490.1 | - | 2 | 792 | |||

Apoe
Genomic Map