siRNA Details
| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009712.3 | NP_033842.3 | 4 | 893 | ||
| GenBank mRNA | AK083309.1 | - | 4 | 877 | ||
| AK140163.1 | - | 4 | 943 | |||
| AK154098.1 | - | 4 | 813 | |||
| AK158312.1 | - | 4 | 894 | |||
| BC150661.1 | - | 4 | 850 | 9087572 | ||
| X92096.1 | - | 3 | 386 | |||

Arsb
Genomic Map